Learn More
Enzo Life Sciences HO 1 polyclonal antibody (1 mg)
SDP
Supplier:  Enzo Life Sciences ADISPA895J
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant rat HO-1 (Hsp32) lacking the membrane spanning region. UniProt: P06762. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Dog. Applications: Flow Cytometry, ICC, IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-676