Learn More
Enzo Life Sciences HO-1 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA894F
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HO-1. UniProt ID: P09601. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Dog. Applications: IHC, WB. Recommended Dilutions: Western Blot (1:1,000, colormetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7583