Learn More
Enzo Life Sciences HO 1 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIOSA150D
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat HO-1. UniProt: P06762. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-009