Learn More
Enzo Life Sciences HO-1 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA894D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HO-1. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Dog. Applications: IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7582