Learn More
Enzo Life Sciences HO 1 polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA896F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1. UniProt: P09601. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Rabbit, Monkey, Hamster, Dog. Applications: IHC (PS), IP, WB. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-678