Learn More
Enzo Life Sciences HO 2 polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA897F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human HO-2. UniProt: P30519. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Porcine, Monkey, Hamster, Dog. Applications: ELISA, ICC, IF, IHC, IP, WB. Recommended dilutions: Immunohistochemistry (1:50). Immunoprecipitation (10µg/ml, denatured). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-681