Learn More
Medchemexpress LLC HY-100453 5mg Medchemexpress, HO-3867 CAS:1172133-28-6 Purity:>98%
SDP
Catalog No. 501931840
Encompass_Preferred
Medchemexpress, HY-100453 5mg HO-3867 CAS:1172133-28-6 HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1840 Supplier Medchemexpress LLC Supplier No. HY1004535MG
Medchemexpress, HY-100453 5mg HO-3867 CAS:1172133-28-6 HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.