Learn More
Selleck Chemical LLC HO-3867
SDP
Supplier:  Selleck Chemical LLC S75015MG
Encompass_Preferred
HO-3867 an analog of curcumin is a selective STAT3 inhibitor that inhibits its phosphorylation transcription and DNA binding without affecting the expression of other active STATs HO-3867 induces apoptosis
Catalog No. 50-217-3280