Learn More
Selleck Chemical LLC HO-3867
Supplier: Selleck Chemical LLC S75011G
HO-3867 an analog of curcumin is a selective STAT3 inhibitor that inhibits its phosphorylation transcription and DNA binding without affecting the expression of other active STATs HO-3867 induces apoptosis