Learn More
Medchemexpress LLC HO-PEG-CH2COOH | 10,000 Da | HO-(CH2CH2O)n-CH2COOH | 1 G
SDP
Catalog No. 5000284803
Encompass_Preferred
HO-PEG-CH2COOH (MW 10,000) is a linear polyethylene glycol polymer terminated with a hydroxyl group and a carboxymethyl (-CH2COOH) group. Supplied as a solid for research use, it is intended as a PEG linker or building block for conjugation chemistries such as PROTAC synthesis and PEGylation.
  • Molecular weight 10,000 Da, providing spacer length for linker design.
  • Hydroxyl and carboxylic acid terminal groups for orthogonal functionalization.
  • Water-soluble polymeric structure suitable for bioconjugation and linker applications.
  • Supplied as a solid with recommended storage at -20°C under inert atmosphere.
  • Available in small research quantities for method development and synthesis.
  • Documentation includes datasheet, COA, and SDS for handling and characterization.
Catalog No. 50-002-84803 Supplier Medchemexpress LLC Supplier No. HYW591338A1G
Add to Cart
Add to Cart
HO-PEG-CH2COOH (MW 10,000) is a linear polyethylene glycol polymer terminated with a hydroxyl group and a carboxymethyl (-CH2COOH) group. Supplied as a solid for research use, it is intended as a PEG linker or building block for conjugation chemistries such as PROTAC synthesis and PEGylation.
  • Molecular weight 10,000 Da, providing spacer length for linker design.
  • Hydroxyl and carboxylic acid terminal groups for orthogonal functionalization.
  • Water-soluble polymeric structure suitable for bioconjugation and linker applications.
  • Supplied as a solid with recommended storage at -20°C under inert atmosphere.
  • Available in small research quantities for method development and synthesis.
  • Documentation includes datasheet, COA, and SDS for handling and characterization.