Learn More
Medchemexpress LLC HO-PEG-CH2COOH | 10000 | HO-(CH2CH2O)n-CH2COOH | 500 MG
SDP
Catalog No. 5000284801
Encompass_Preferred
HO-PEG-CH2COOH (MW 10,000) is a polyethylene glycol (PEG) polymer terminated with a hydroxyl group and a terminal carboxylic acid. The manufacturer describes it as suitable for use as a PROTAC linker in the synthesis of PROTAC molecules and supplies product documentation including a data sheet and SDS.
Catalog No. 50-002-84801 Supplier Medchemexpress LLC Supplier No. HYW591338A500MG
HO-PEG-CH2COOH (MW 10,000) is a polyethylene glycol (PEG) polymer terminated with a hydroxyl group and a terminal carboxylic acid. The manufacturer describes it as suitable for use as a PROTAC linker in the synthesis of PROTAC molecules and supplies product documentation including a data sheet and SDS.