Learn More
Medchemexpress LLC HO-PEG13-OH | 17598-96-8 | ≥98.0% | C26H54O14 | 250 MG
SDP
Catalog No. 5000352897
Encompass_Preferred
HO-PEG13-OH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is a white to off-white solid with a molecular weight of 590.70. This compound is designed for research purposes, facilitating the creation of PROTACs that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid
Catalog No. 50-003-52897 Supplier Medchemexpress LLC Supplier No. 250MGHY141234
HO-PEG13-OH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is a white to off-white solid with a molecular weight of 590.70. This compound is designed for research purposes, facilitating the creation of PROTACs that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid