Learn More
Medchemexpress LLC HO-PEG20-OH | 351342-08-0 | 98.22% | C40H82O21 | 250 MG
SDP
Catalog No. 5000352883
Encompass_Preferred
HO-PEG20-OH is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52883 Supplier Medchemexpress LLC Supplier No. 250MGHY141236
HO-PEG20-OH is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system
  • Selectively degrades target proteins