Learn More
Medchemexpress LLC Ho-peg24-oh | 2243942-52-9 | 99.9% | C48H98O25 | 100 MG
SDP
Catalog No. 5000354344
Encompass_Preferred
This product is a PEG-based linker designed for use in the synthesis of PROTACs. These small molecules represent an innovative approach to developing targeted therapeutic agents. The linker's structure is instrumental in creating advanced drug candidates, enabling the formation of PROTACs for various research applications.
  • Functions as a PEG-based linker.
  • Essential for the synthesis of PROTACs.
  • High purity, reaching 99.89%.
  • Exhibits solubility in DMSO.
  • Stable as a powder for up to 3 years at -20°C and 2 years at 4°C; in solvent, stable for 6 months at -80°C and 1 month at -20°C.
Catalog No. 50-003-54344 Supplier Medchemexpress LLC Supplier No. HY141238100MG
This product is a PEG-based linker designed for use in the synthesis of PROTACs. These small molecules represent an innovative approach to developing targeted therapeutic agents. The linker's structure is instrumental in creating advanced drug candidates, enabling the formation of PROTACs for various research applications.
  • Functions as a PEG-based linker.
  • Essential for the synthesis of PROTACs.
  • High purity, reaching 99.89%.
  • Exhibits solubility in DMSO.
  • Stable as a powder for up to 3 years at -20°C and 2 years at 4°C; in solvent, stable for 6 months at -80°C and 1 month at -20°C.