Learn More
Medchemexpress LLC HO-PEG24-OH | 2243942-52-9 | 99.9% | C48H98O25 | 50 MG
SDP
Catalog No. 5000353285
Encompass_Preferred
HO-PEG24-OH is a high-purity, polyethylene glycol (PEG)-based PROTAC linker with a purity of 99.89%. This linker is specifically designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs), providing a versatile building block for targeted protein degradation research.
  • Peg-based linker
  • Designed for PROTAC synthesis
  • High purity for research applications
  • Supports targeted protein degradation studies
Catalog No. 50-003-53285 Supplier Medchemexpress LLC Supplier No. HY14123850MG
HO-PEG24-OH is a high-purity, polyethylene glycol (PEG)-based PROTAC linker with a purity of 99.89%. This linker is specifically designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs), providing a versatile building block for targeted protein degradation research.
  • Peg-based linker
  • Designed for PROTAC synthesis
  • High purity for research applications
  • Supports targeted protein degradation studies