Learn More
Medchemexpress LLC HO-PEG3-(CH2)6-Cl | 1355955-95-1 | MFCD32705069 | 98.0% | 268.78 g/mol | C12H25ClO4 | 25 MG
SDP
Catalog No. 5000284394
Encompass_Preferred
HO-PEG3-(CH2)6-Cl is a PEG3-based PROTAC linker containing a six-carbon chloroalkyl spacer and a terminal hydroxyl. It is supplied as a small-mass research chemical for use in chemical biology and targeted protein degradation research.
  • PEG3 spacer with flexible ether linkages.
  • Terminal hydroxyl and chloro groups for orthogonal conjugation.
  • Molecular formula C12H25ClO4 and molecular weight 268.78 g/mol.
  • Provided as a 25 mg research sample.
  • Intended for research use; consult product documentation for storage and purity.
Catalog No. 50-002-84394 Supplier Medchemexpress LLC Supplier No. HYW61360725MG
HO-PEG3-(CH2)6-Cl is a PEG3-based PROTAC linker containing a six-carbon chloroalkyl spacer and a terminal hydroxyl. It is supplied as a small-mass research chemical for use in chemical biology and targeted protein degradation research.
  • PEG3 spacer with flexible ether linkages.
  • Terminal hydroxyl and chloro groups for orthogonal conjugation.
  • Molecular formula C12H25ClO4 and molecular weight 268.78 g/mol.
  • Provided as a 25 mg research sample.
  • Intended for research use; consult product documentation for storage and purity.