Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ho-peg5k-nhs

Catalog No. 503125527 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5527 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-5527 Supplier Sigma Aldrich Supplier No. JKA50781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydroxyl-PEG-NHS Ester (HO-PEG-NHS) is hydrophilic in nature.HO-PEG-NHS may be conjugated on to a hydrophobic H40-PLA-COOH molecule to form H40-PLA-PEG-Apt) (aptamer-conjugated). Other applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization.Product of JenKem Technology

Specifications

Linear Formula HO(CH2CH2O)nCH2COOC4H2O2N
Quantity 1 g
Synonym HO-PEG-NHS; Hydroxyl-PEG-NHS Ester
Molecular Weight (g/mol) 5000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.