Learn More
Bioworld Hoagland Medium, Nitrogen-Free, 100 L
Catalog No. 50254629
Encompass_Preferred
Hoagland MediumContains Macronutrients and Micronutrients for watering, nitrogen fixing plants grown in vermiculiteNitrogen freeSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, 50L, or 100L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedUse without autoclavingUse 1.34 g/L
Catalog No. 50-254-629 Supplier Bioworld Supplier No. 306300385
Hoagland MediumContains Macronutrients and Micronutrients for watering, nitrogen fixing plants grown in vermiculiteNitrogen freeSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, 50L, or 100L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedUse without autoclavingUse 1.34 g/L