Learn More
Apexbio Technology LLC HOAt, 100g. Cas: 39968-33-7 MFCD: MFCD00210053. coupling activator for racemization-free coupling in peptide synthesis
SDP
Supplier:  Apexbio Technology LLC A7024100000
Encompass_Preferred
A coupling activator for racemization-free coupling in peptide synthesis; An additive for peptide segment coupling in in reducing the extent of configurational loss at the reactive carboxylic acid residue. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4969