Learn More
Apexbio Technology LLC HOAt 39968-33-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061486
Encompass_Preferred
HOAt (1-Hydroxy-7-azabenzotriazole) is a peptide coupling reagent frequently employed in peptide synthesis for the activation of carboxylic groups Structurally HOAt serves as an activating additive that forms reactive intermediates facilitating amide bond formation while mitigating side reactions such as racemization As reported in experimental peptide coupling studies combining HOAt with diisopropylcarbodiimide (DIC) promotes coupling reactions with improved retention of stereochemical integrity relative to traditional coupling systems utilizing other benzotriazole derivatives HOAt is thus commonly used both in segment coupling and stepwise solid-phase peptide assembly methodologies aimed at synthesizing peptides requiring minimal epimerization
Catalog No. 50-000-61486 Supplier Apexbio Technology LLC Supplier No. A70245.1
HOAt (1-Hydroxy-7-azabenzotriazole) is a peptide coupling reagent frequently employed in peptide synthesis for the activation of carboxylic groups Structurally HOAt serves as an activating additive that forms reactive intermediates facilitating amide bond formation while mitigating side reactions such as racemization As reported in experimental peptide coupling studies combining HOAt with diisopropylcarbodiimide (DIC) promotes coupling reactions with improved retention of stereochemical integrity relative to traditional coupling systems utilizing other benzotriazole derivatives HOAt is thus commonly used both in segment coupling and stepwise solid-phase peptide assembly methodologies aimed at synthesizing peptides requiring minimal epimerization