Learn More
Onset Computer Corporation HOBO 5-INCH PROBE TEMPERATURE
SDP
Catalog No. NC1402285
Encompass
The HOBO U12 Stainless 5-inch Probe Temperature Data Logger is ideal for monitoring internal temperature of foods or any application where fast response is critical.Highlighted Features Food-grade 316 Series Stainless Steel High-accuracy, extended temperature range Suitable for high pressure applications to 2200psi Fast readout via direct USB interface Compatible with HOBOware and HOBOware Pro software for logger setup, graphing and analysis
Catalog No. NC1402285 Supplier Onset Computer Corporation Supplier No. U1201502
The HOBO U12 Stainless 5-inch Probe Temperature Data Logger is ideal for monitoring internal temperature of foods or any application where fast response is critical.Highlighted Features Food-grade 316 Series Stainless Steel High-accuracy, extended temperature range Suitable for high pressure applications to 2200psi Fast readout via direct USB interface Compatible with HOBOware and HOBOware Pro software for logger setup, graphing and analysis