Learn More
Microdaq Onset HOBO MX1101 Bluetooth Temperature and Humidity Data Logger
SDP
Catalog No. NC1108657
Encompass
Integrated Sensors for Temperature and Humidity Measurements in an Indoor Environment. Wireless Transmission of Recorded Readings to Android devices, iPhone, iPad Using Bluetooth Smart LE Communications. -20°C to 70°C and 1% to 90% RH Measurement Ranges with a ±0.21°C and 2% RH Accuracy. (2) User-Replaceable AAA Alkaline Batteries with 1 Year Life (1 Minute Sampling Rate). Stores up to 84,650 Measured Readings with Date/Time Stamp if Bluetooth Communications Not Available. Includes Command Strip, Double-Sided Tape, Hook & Loop Strap and (2) AAA Alkaline Batteries. HOBOmobile App Available on the Apple Store and the Google Play Store.
Catalog No. NC1108657 Supplier Microdaq Supplier No. MX1101
Integrated Sensors for Temperature and Humidity Measurements in an Indoor Environment. Wireless Transmission of Recorded Readings to Android devices, iPhone, iPad Using Bluetooth Smart LE Communications. -20°C to 70°C and 1% to 90% RH Measurement Ranges with a ±0.21°C and 2% RH Accuracy. (2) User-Replaceable AAA Alkaline Batteries with 1 Year Life (1 Minute Sampling Rate). Stores up to 84,650 Measured Readings with Date/Time Stamp if Bluetooth Communications Not Available. Includes Command Strip, Double-Sided Tape, Hook & Loop Strap and (2) AAA Alkaline Batteries. HOBOmobile App Available on the Apple Store and the Google Play Store.