Learn More
Onset Computer Corporation HOBO Pendant Temperature/Light 64K Data Logger
SDP
Catalog No. NC1643979
Encompass
This miniature data logger can record temperature and relative light levels. Complete with waterproof casing, this product is designed for indoor, outdoor, and underwater deployment. This model can store approximately 52,000 measurments of 10-bit readings. Use a solar radiation shield for accurate temperature measurement in sunlight.
Catalog No. NC1643979 Supplier Onset Computer Corporation Supplier No. UA00264
This miniature data logger can record temperature and relative light levels. Complete with waterproof casing, this product is designed for indoor, outdoor, and underwater deployment. This model can store approximately 52,000 measurments of 10-bit readings. Use a solar radiation shield for accurate temperature measurement in sunlight.