Learn More
Apexbio Technology LLC HOBt (anhydrous), 500g. Cas: 2592-95-2 MFCD: MFCD00005805. Racemization inhibitor
SDP
Catalog No. 501904971
Encompass_Preferred
Hydroxy benzotriazole, abbreviated HOBt, is anorganic compoundthat is a derivative of benzotriazole. HOBt, as a commercial product, is a white crystalline powder contains some water (~11.7% wt as the HOBt monohydrate crystal). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4971 Supplier Apexbio Technology LLC Supplier No. A7025500000
Hydroxy benzotriazole, abbreviated HOBt, is anorganic compoundthat is a derivative of benzotriazole. HOBt, as a commercial product, is a white crystalline powder contains some water (~11.7% wt as the HOBt monohydrate crystal). Other sizes are available. Please inqury us for quote.