Learn More
StatLab Hodgkins Lymphoma IHC Control Slides
SDP
Catalog No. 21033338
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
21-033-338 25 Pk.
1 options
Catalog No. 21-033-338 Supplier StatLab Supplier No. CSH1025P
Add to Cart
Add to Cart
Use hodgkins lymphoma IHC control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.