Learn More
Cayman Chemical Hodgkinsine, Purity: ≥95%, CAS: 18210-71-4, 5 mg
SDP
Catalog No. 502991124
Encompass_Preferred
An alkaloid with analgesic activity; increases latency to paw licking in the hot plate test and latency to tail withdrawal in the tail-flick test in mice at 5 mg/kg
Catalog No. 50-299-1124 Supplier Cayman Chemical Supplier No. 314135 MG
An alkaloid with analgesic activity; increases latency to paw licking in the hot plate test and latency to tail withdrawal in the tail-flick test in mice at 5 mg/kg