Learn More
Enzo Life Sciences HOE-140 (1mg). CAS: 130308-48-4
SDP
Supplier:  Enzo Life Sciences BMLNK1040001
Encompass_Preferred
A potent and selective bradykinin B2 antagonist. HOE-140 is a useful tool for discrimination of the physiological roles of bradykinin B2 versus B1 receptors. Displays a protective effect in myocardial reperfusion injury. Alternative name: Icatibant. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1198