Learn More
CPC Scientific H-DArg-Arg-Pro-Hyp-Gly-Thi-Ser-DTic-Oic-Arg-OH 1MG
SDP
Catalog No. 502108850
Encompass
Bradykinin B2 receptor antagonist that prevents the bradykinin-induced incapacitation.SEQUENCE: H-DArg-Arg-Pro-Hyp-Gly-Thi-Ser-DTic-Oic-Arg-OHONE-LETTER SEQUENCE: DArg-RP-Hyp-G-Thi-S-DTic-Oic-RMOLECULAR FORMULA: C59H89N19O13S1MOLECULAR WEIGHT: 1304.52STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [130308-48-4]SYNONYMS: Icatibant, Firazyr, CS-1399, HOE-140, HOE140RESEARCH AREA: CardiovascularREFERENCES:C. R. Tonussi et al., European Journal of Pharmacology, 326, 61 (1997)SKU(s): BRAK-003A, BRAK-003B
Catalog No. 50-210-8850 Supplier CPC Scientific Supplier No. BRAK003B
Bradykinin B2 receptor antagonist that prevents the bradykinin-induced incapacitation.SEQUENCE: H-DArg-Arg-Pro-Hyp-Gly-Thi-Ser-DTic-Oic-Arg-OHONE-LETTER SEQUENCE: DArg-RP-Hyp-G-Thi-S-DTic-Oic-RMOLECULAR FORMULA: C59H89N19O13S1MOLECULAR WEIGHT: 1304.52STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [130308-48-4]SYNONYMS: Icatibant, Firazyr, CS-1399, HOE-140, HOE140RESEARCH AREA: CardiovascularREFERENCES:C. R. Tonussi et al., European Journal of Pharmacology, 326, 61 (1997)SKU(s): BRAK-003A, BRAK-003B