Learn More
Medchemexpress LLC HY-15561 5mg Medchemexpress, HOE-S 785026 CAS:132869-83-1 Purity:>98%
SDP
Catalog No. 502081735
Encompass_Preferred
Medchemexpress, HY-15561 5mg HOE-S 785026 CAS:132869-83-1 HOE-S 785026 is a blue fluorescent dyes, which can be used as a cell dye for DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1735 Supplier Medchemexpress LLC Supplier No. HY155615MG
Medchemexpress, HY-15561 5mg HOE-S 785026 CAS:132869-83-1 HOE-S 785026 is a blue fluorescent dyes, which can be used as a cell dye for DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.