Learn More
Biotium Hoechst 33258
Catalog No. 501964644
Encompass_Preferred
Hoechst dyes are cell membrane-permeant, minor groove-binding blue fluorescent DNA stains. These dyes are widely used in cell cycle and apoptosis studies as nuclear counterstains. Hoechst 33528 is slightly more water soluble than Hoechst 33342. The dyes can be used to stain live or fixed cells in buffer or medium at 1 ug/mL, with no wash step required. Properties: Yellow solid; Store dessicated at -20°C, protect from light; Ex/Em= 352/461 nm (with DNA); C25H27Cl3N6O; MW: 623.96; [23491-45-4].
Catalog No. 50-196-4644 Supplier Biotium Supplier No. 40045
Hoechst dyes are cell membrane-permeant, minor groove-binding blue fluorescent DNA stains. These dyes are widely used in cell cycle and apoptosis studies as nuclear counterstains. Hoechst 33528 is slightly more water soluble than Hoechst 33342. The dyes can be used to stain live or fixed cells in buffer or medium at 1 ug/mL, with no wash step required. Properties: Yellow solid; Store dessicated at -20°C, protect from light; Ex/Em= 352/461 nm (with DNA); C25H27Cl3N6O; MW: 623.96; [23491-45-4].