Learn More
Cayman Chemical Hoechst 33258, Purity: ≥95%, CAS: 23491-45-4, 250 mg
SDP
Catalog No. 502964932
Encompass_Preferred
A cell-permeable, fluorescent probe for DNA (ex. 352 nm/em. max 461 nm) that is useful as a marker of nuclei for cell cycle and to distinguish nuclear morphology in apoptotic cells
Catalog No. 50-296-4932 Supplier Cayman Chemical Supplier No. 16756250 MG
A cell-permeable, fluorescent probe for DNA (ex. 352 nm/em. max 461 nm) that is useful as a marker of nuclei for cell cycle and to distinguish nuclear morphology in apoptotic cells