Learn More
AdipoGen Hoechst 33258
SDP
Supplier:  AdipoGen CDXB0029G005
Encompass_Preferred
Chemical. CAS 23491-45-4. Formula C25H24N6O . 3HCl. MW 533.88. Synthetic. Useful for staining DNA, chromosomes and nuclei. Binds to the minor groove of DNA at AT-rich sequences. This dye is commonly used for determining the DNA content of viable cells without detergent treatment or fixation and for fluorescence microscopy or flow cytometry.
Catalog No. 50-256-1703