Learn More
Aat Bioquest Hoechst 33258 CAS 23491-45-4 (100 mg)
SDP
Catalog No. 502249726
Encompass_Preferred
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 100 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9726 Supplier Aat Bioquest Supplier No. 17520
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 100 mg.