Learn More
Aat Bioquest Hoechst 33258 20 mM solution in water (5 mL)
SDP
Supplier:  Aat Bioquest 17525
Encompass_Preferred
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 5 mL.
Catalog No. 50-224-9728