Learn More
Novus Biologicals™ Hoechst 33258 Inhibitor

Description
Biological Activity: Fluorescent dye. Binds the minor groove of DNA at A and T rich regions. Can be used as indicator of apoptosis.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 23491-45-4 |
| Formulation | C25H24N6O3.HCl |
| For Use With (Application) | Staining solution for DNA |
| Quantity | 50 mg |
| Product Type | Staining solution |
| Molecular Weight (g/mol) | 533.33g/mol |
| Solubility | Soluble to 20mM in DMSO,100mM in water |
| Content And Storage | Room Temperature |
| Color | Yellow |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.