Learn More
AnaSpec Hoechst 33342, 20 mM solution in water, | MW 561.93 | (5 mL)
Catalog No. 50850587
Encompass
Hoechst 33342, 20 mM solution in water, ; MW 561.93; (5 mL) Cell-permeant DNA minor groove binding dye
Catalog No. 50-850-587 Supplier AnaSpec Supplier No. AS83218
Hoechst 33342, 20 mM solution in water, ; MW 561.93; (5 mL) Cell-permeant DNA minor groove binding dye