Learn More
Aat Bioquest Hoechst 33342 Ultrapure Grade (100 mg)
SDP
Catalog No. 502249729
Encompass_Preferred
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 100 mg.
Catalog No. 50-224-9729 Supplier Aat Bioquest Supplier No. 17530
Add to Cart
Add to Cart
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 100 mg.