Learn More
Aat Bioquest Hoechst 33342 20 mM solution in water (5 mL)
SDP
Catalog No. 502249731
Encompass_Preferred
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 5 mL.
This item is not returnable. View return policy
Catalog No. 50-224-9731 Supplier Aat Bioquest Supplier No. 17535
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy. This catalog is for 5 mL.