Learn More
Aat Bioquest Hoechst 33342 NHS ester 1 mg
SDP
Catalog No. NC3404285
Encompass_Preferred
Hoechst 33342 NHS ester is an excellent building block for developing site-specific nucleic acid probes. It can be readily conjugated to amino-containing biomolecules (such as antibodies, peptides and amino-modified oligos). Hoechst 33342 is a DNA-specific dye that binds to the minor groove of double-stranded DNA. It emits blue fluorescence when bound to DNA. Its excitation maximum is around 350 nm, and the emission maximum is approximately 460 nm. Its ability to readily penetrate live cell membranes makes it suitable for staining live cells. This property allows researchers to monitor nuclear morphology and DNA content in living cells. Hoechst 33342 is commonly used in fluorescence microscopy to visualize the nucleus of cells. It selectively stains the DNA in the cell nucleus, and is often used in combination with other fluorochromes to study cell structure and dynamics. Hoechst 33342 is used for cell cycle analysis in flow cytometry.
Catalog No. NC3404285 Supplier Aat Bioquest Supplier No. 17670
Hoechst 33342 NHS ester is an excellent building block for developing site-specific nucleic acid probes. It can be readily conjugated to amino-containing biomolecules (such as antibodies, peptides and amino-modified oligos). Hoechst 33342 is a DNA-specific dye that binds to the minor groove of double-stranded DNA. It emits blue fluorescence when bound to DNA. Its excitation maximum is around 350 nm, and the emission maximum is approximately 460 nm. Its ability to readily penetrate live cell membranes makes it suitable for staining live cells. This property allows researchers to monitor nuclear morphology and DNA content in living cells. Hoechst 33342 is commonly used in fluorescence microscopy to visualize the nucleus of cells. It selectively stains the DNA in the cell nucleus, and is often used in combination with other fluorochromes to study cell structure and dynamics. Hoechst 33342 is used for cell cycle analysis in flow cytometry.