Learn More
Apexbio Technology LLC Hoechst 33342/PI Double Staining Kit 100T
SDP
Catalog No. 5000097290
Encompass_Preferred
Hoechst 33342 PI Double Staining Kit is a fluorescence-based assay kit designed to distinguish apoptosis and necrosis in cell populations The kit utilizes two fluorescent probes Hoechst 33342 and propidium iodide (PI) to assess plasma membrane integrity and nuclear morphology Hoechst 33342 penetrates live cells and binds to DNA producing blue fluorescence with intensity increasing upon apoptotic chromatin condensation PI is impermeable to intact cell membranes and selectively labels necrotic cells with red fluorescence Compared to single-stain apoptosis assays this dual-stain approach enables simultaneous detection of viable apoptotic and necrotic cells Based on these properties Hoechst 33342 PI Double Staining Kit is applicable in pharmacological evaluation and cytotoxicity studies
Catalog No. 50-000-97290 Supplier Apexbio Technology LLC Supplier No. K2237100
Add to Cart
Add to Cart
Hoechst 33342 PI Double Staining Kit is a fluorescence-based assay kit designed to distinguish apoptosis and necrosis in cell populations The kit utilizes two fluorescent probes Hoechst 33342 and propidium iodide (PI) to assess plasma membrane integrity and nuclear morphology Hoechst 33342 penetrates live cells and binds to DNA producing blue fluorescence with intensity increasing upon apoptotic chromatin condensation PI is impermeable to intact cell membranes and selectively labels necrotic cells with red fluorescence Compared to single-stain apoptosis assays this dual-stain approach enables simultaneous detection of viable apoptotic and necrotic cells Based on these properties Hoechst 33342 PI Double Staining Kit is applicable in pharmacological evaluation and cytotoxicity studies