Learn More
Medchemexpress LLC HY-15559A 100mg , Hoechst 33342 (trihydrochloride) CAS:875756-97-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15559A100MG
Encompass_Preferred
HY-15559A 100mg Hoechst 33342 (trihydrochloride) CAS: 875756-97-1 Hoechst 33342 trihydrochloride is a membrane permeant blue fluorescent DNA stain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9754