Learn More
Enzo Life Sciences Hoechst 33342 (ultra pure) (100mg). CAS: 23491-52-3
SDP
Catalog No. 502012636
Encompass_Preferred
Hoechst 33342 is fluorescent probe useful for detecting DNA by fluorescence microscopy and flow cytometry. Hoechst 33342 may be used on live or fixed cells and is also applicable for cell cycle analysis and monitoring DNA condensation by flow cytometry. An additional ethyl group renders Hoechst 33342 more lipophilic, and thus more readily cell permeable than Hoechst 33258 . Exclusion of Hoechst 33342 dye is a characteristic common to stem cells, as well as chemotherapy-resistant cancer cells. Wavelength Maxima: Excitation 350nm, Emission 461nm Alternative name: 2'-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bi-1H-benzimidazole. Purity: ≥95% (HPLC). Solubility: Soluble in water (50mg/ml) or DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2636 Supplier Enzo Life Sciences Supplier No. ENZ52401
Hoechst 33342 is fluorescent probe useful for detecting DNA by fluorescence microscopy and flow cytometry. Hoechst 33342 may be used on live or fixed cells and is also applicable for cell cycle analysis and monitoring DNA condensation by flow cytometry. An additional ethyl group renders Hoechst 33342 more lipophilic, and thus more readily cell permeable than Hoechst 33258 . Exclusion of Hoechst 33342 dye is a characteristic common to stem cells, as well as chemotherapy-resistant cancer cells. Wavelength Maxima: Excitation 350nm, Emission 461nm Alternative name: 2'-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bi-1H-benzimidazole. Purity: ≥95% (HPLC). Solubility: Soluble in water (50mg/ml) or DMSO (10mg/ml). Long Term Storage: -20°C.