Learn More
Cayman Chemical Hoechst 33342 (hydrochloride) (solution), Purity: ≥98%, CAS: 875756-97-1, 2 mL
SDP
Catalog No. 502945371
Encompass_Preferred
A fluorescent DNA probe; forms a fluorescent complex by binding to the minor groove of adenine and thymine-rich sequences; displays an emission maximum of 465 when bound to dsDNA upon excitation at 350 nm; has been used as a marker of nuclei in cell cycle studies and to distinguish nuclear morphology in apoptotic cells
Catalog No. 50-294-5371 Supplier Cayman Chemical Supplier No. 407972 ML
A fluorescent DNA probe; forms a fluorescent complex by binding to the minor groove of adenine and thymine-rich sequences; displays an emission maximum of 465 when bound to dsDNA upon excitation at 350 nm; has been used as a marker of nuclei in cell cycle studies and to distinguish nuclear morphology in apoptotic cells