Learn More
Cayman Chemical Hoechst 33342hydrochlorIde 5mL
SDP
Catalog No. 502942547
Encompass_Preferred
A fluorescent DNA probe; forms a fluorescent complex by binding to the minor groove of adenine and thymine-rich sequences; displays an emission maximum of 465 when bound to dsDNA upon excitation at 350 nm; has been used as a marker of nuclei in cell cycle studies and to distinguish nuclear morphology in apoptotic cells
Catalog No. 50-294-2547 Supplier Cayman Chemical Supplier No. 407975 ML
A fluorescent DNA probe; forms a fluorescent complex by binding to the minor groove of adenine and thymine-rich sequences; displays an emission maximum of 465 when bound to dsDNA upon excitation at 350 nm; has been used as a marker of nuclei in cell cycle studies and to distinguish nuclear morphology in apoptotic cells