Learn More
Cayman Chemical Hoechst 33342 (hydrochloride), Purity: ≥98%, CAS: 875756-97-1, 50 mg
SDP
Catalog No. 502973345
Encompass_Preferred
A cell-permeable, fluorescent probe for DNA that is useful as a marker of nuclei for cell cycle studies in combination with BrdU and to distinguish nuclear morphology in apoptotic cells
This item is not returnable. View return policy
Catalog No. 50-297-3345 Supplier Cayman Chemical Supplier No. 1554750 MG
A cell-permeable, fluorescent probe for DNA that is useful as a marker of nuclei for cell cycle studies in combination with BrdU and to distinguish nuclear morphology in apoptotic cells