Learn More
Cayman Chemical Hoechst 33342 (hydrochloride), Purity: ≥98%, CAS: 875756-97-1, 500 mg
SDP
Catalog No. 502973343
Encompass_Preferred
A cell-permeable, fluorescent probe for DNA that is useful as a marker of nuclei for cell cycle studies in combination with BrdU and to distinguish nuclear morphology in apoptotic cells
Catalog No. 50-297-3343 Supplier Cayman Chemical Supplier No. 15547500 MG
A cell-permeable, fluorescent probe for DNA that is useful as a marker of nuclei for cell cycle studies in combination with BrdU and to distinguish nuclear morphology in apoptotic cells