Learn More
Apexbio Technology LLC Hoechst 34580 23555-00-2 100mg
SDP
Catalog No. 5000063228
Encompass_Preferred
Hoechst 34580 is a blue-fluorescent dye routinely employed in biomedical research for labeling cell nuclei and analyzing cell-cycle progression This dye binds selectively to nucleic acids predominantly by interacting with DNA via minor groove binding allowing visualization of nuclear morphology and chromatin structure Upon binding DNA Hoechst 34580 exhibits fluorescence excitation around 392 nm and emission maxima near 440 460 nm suitable for standard fluorescent microscopy Typical working concentrations for cell labeling range between 0 5 and 10 g/mL Due to these characteristics Hoechst 34580 is widely used for fluorescence microscopy flow cytometry nuclear staining applications and monitoring cellular proliferation in biological investigations
Catalog No. 50-000-63228 Supplier Apexbio Technology LLC Supplier No. A3475100
Hoechst 34580 is a blue-fluorescent dye routinely employed in biomedical research for labeling cell nuclei and analyzing cell-cycle progression This dye binds selectively to nucleic acids predominantly by interacting with DNA via minor groove binding allowing visualization of nuclear morphology and chromatin structure Upon binding DNA Hoechst 34580 exhibits fluorescence excitation around 392 nm and emission maxima near 440 460 nm suitable for standard fluorescent microscopy Typical working concentrations for cell labeling range between 0 5 and 10 g/mL Due to these characteristics Hoechst 34580 is widely used for fluorescence microscopy flow cytometry nuclear staining applications and monitoring cellular proliferation in biological investigations