Learn More
Medchemexpress LLC HY-15560B 5mg Medchemexpress, Hoechst 34580 (tetrahydrochloride) CAS: Purity:>98%
SDP
Catalog No. 502081734
Encompass_Preferred
Medchemexpress, HY-15560B 5mg Hoechst 34580 (tetrahydrochloride) CAS: Hoechst 34580 tetrahydrochloride is a cell-permeable fluorescent dye for staining DNA and nuclei. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1734 Supplier Medchemexpress LLC Supplier No. HY15560B5MG
Medchemexpress, HY-15560B 5mg Hoechst 34580 (tetrahydrochloride) CAS: Hoechst 34580 tetrahydrochloride is a cell-permeable fluorescent dye for staining DNA and nuclei. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.