Learn More
Hoefer, Inc Hoefer Inc. Autocap Dispenser, 6-Channel, 8-Channel, 1 ea
SDP
Catalog No. NC3942891
Encompass_Preferred
The Autocap Dispenser is a fully automated solution designed for the efficient dispensing of cells, reference standards, and reagents. It supports 6-channel and 8-channel configurations to simultaneously decap, recap, and dispense into a wide variety of sample tubes, streamlining laboratory workflows with high consistency and precision.
  • Throughput of 2000 sample tubes per hour
  • Filling range from 50 μL to 2 mL
  • Dispensing volume error of less than 5%
  • High dispensing precision within 0.02 mm
  • Enhanced safety through disposable sterile consumables
  • Integrated UV sterilization lamps for module disinfection
  • Compact, portable cabinet suitable for biosafety cabinets
  • Minimal exposure time of 10 seconds per cycle
Catalog No. NC3942891 Supplier Hoefer, Inc Supplier No. AUTOCAP DISPENSER
The Autocap Dispenser is a fully automated solution designed for the efficient dispensing of cells, reference standards, and reagents. It supports 6-channel and 8-channel configurations to simultaneously decap, recap, and dispense into a wide variety of sample tubes, streamlining laboratory workflows with high consistency and precision.
  • Throughput of 2000 sample tubes per hour
  • Filling range from 50 μL to 2 mL
  • Dispensing volume error of less than 5%
  • High dispensing precision within 0.02 mm
  • Enhanced safety through disposable sterile consumables
  • Integrated UV sterilization lamps for module disinfection
  • Compact, portable cabinet suitable for biosafety cabinets
  • Minimal exposure time of 10 seconds per cycle