Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Coomassie Brilliant Blue G-250
SDP

Catalog No. 03500517
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-500-517 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-517 Supplier Hoefer™ Supplier No. GR13425
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Assay >99.9%

  • Dark blue/purple crystalline powder

Specifications

Color Brilliant Blue
Quantity 25 g
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.